missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hexafluoroantimonate(V), 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 500 g |
| UN Number | UN1549 |
| Formula Weight | 258.73 |
| Percent Purity | 98% |
| Odor | Odorless |
| Density | 3.38 |
| Chemical Name or Material | Sodium hexafluoroantimonate(V) |
Safety and Handling
| Hazard Category | H302+H332 |
| Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S. |
| EINECSNumber | 240-989-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only