missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexafluoroantimonate, 98%

Catalog No. AC419540250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419540250 25 g Plastic bottle
AC419541000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC419540250 Supplier Thermo Scientific Chemicals Supplier No. 419540250
Only null left

CAS: 16925-25-0 | F6NaSb | 258.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 16925-25-0
Molecular Formula F6NaSb
Molecular Weight (g/mol) 258.72
MDL Number MFCD00003482
InChI Key HKLMYZVMEYYVBS-UHFFFAOYSA-H
Synonym sodium hexafluoroantimonate, sodium hexafluoroantimonate v, sodium hexafluorostibanuide, sodium hexafluoro antimonate, antimony sodium fluoride, f6sb.na, nasbf6, acmc-20ajo7, sodium hexafluorostiboranuide, hexafluorostibine, sodium salt
PubChem CID 16689647
IUPAC Name sodium;hexafluoroantimony(1-)
SMILES F[Sb-](F)(F)(F)(F)F.[Na+]

Specifications

Color White
Physical Form Fine Crystalline Powder
Quantity 25 g
Assay Percent Range 97.5% min
Linear Formula NaSbF6
Solubility Information Solubility in water: very soluble
Formula Weight 258.72
Percent Purity 98%
Packaging Plastic bottle
Chemical Name or Material Sodium hexafluoroantimonate, 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 240-989-8
TSCA TSCA

RUO – Research Use Only