missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hexadecyl Sulfate (contains ca. 40% Sodium Stearyl Sulfate) 95.0+%, TCI America™

Catalog No. H0080500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0080500G 500 g
H008025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0080500G Supplier TCI America Supplier No. H0080500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1120-01-0
Molecular Formula C16H33NaO3S
Molecular Weight (g/mol) 328.49
MDL Number MFCD00047766
InChI Key PNGBYKXZVCIZRN-UHFFFAOYSA-M
Synonym sodium hexadecyl sulfate, sodium cetyl sulfate, sodium n-hexadecyl sulfate, unii-3v3y3o7biq, conco sulfate c, avitex c, avitex sf, sodium hexyldecyl sulfate, tergitol anionic 7, 3v3y3o7biq
PubChem CID 23695542
IUPAC Name sodium hexadecane-1-sulfonate
SMILES [Na+].CCCCCCCCCCCCCCCCS([O-])(=O)=O

Specifications

Color White
Quantity 500 g
Formula Weight 344.49
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Hexadecyl Sulfate (contains ca. 40% Sodium Stearyl Sulfate)
EINECSNumber (2)-1679
RTECSNumber MM0225700
TSCA Yes