missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexachloroplatinate(IV) hexahydrate

Catalog No. AA1104703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1104703 1 g
AA1104706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1104703 Supplier Thermo Scientific Chemicals Supplier No. 01104703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Use As a Catalyst | CAS: 19583-77-8 | Cl6H12Na2O6Pt | 561.85 g/mol

Catalyst.Sodium hexachloroplatinate(IV) hexahydrate is used as a catalyst. It acts as a source of platinum in the preparation of platinum complex. It is also used in the synthesis of platinum nanoparticles using glutathione as a reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19583-77-8
Molecular Formula Cl6H12Na2O6Pt
Molecular Weight (g/mol) 561.85
MDL Number MFCD00149172
InChI Key XGFSWVNCHVSCSP-UHFFFAOYSA-H
Synonym sodium hexachloroplatinate iv hexahydrate, cl6pt.2na.6h2o, platinum iv sodium chloride, disodium;hexachloroplatinum 2-;hexahydrate, disodium hexachloroplatinate 2-hexahydrate
PubChem CID 24846071
SMILES O.O.O.O.O.O.[Na+].[Na+].Cl[Pt](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 110°C-6H2O
Color Orange
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water,alcohol. Insoluble in ether.
IUPAC Name disodium;hexachloroplatinum(2-);hexahydrate
Formula Weight 561.88 (453.79 Anhydrous)
Odor Odorless
Appearance Orange powder
Density 2.5
Chemical Name or Material Sodium hexachloroplatinate(IV) hexahydrate
Show More Show Less
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 240-983-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only