missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexachloroplatinate de sodium(IV) hexahydraté, 98%
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Spécifications
Spécifications
| Color | Orange |
| Physical Form | Solide cristallin |
| Quantity | 1 g |
| Assay Percent Range | 98% |
| Linear Formula | Na2PtCl6·6H2O |
| Solubility Information | Solubility in water: soluble in water |
| Formula Weight | 561.88 |
| Percent Purity | 98% |
| Packaging | Bouteille en verre |
| Chemical Name or Material | Sodium hexachloroplatinate(IV) hexahydrate, 98% |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. Wear eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Usage réservé à la recherche