missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexabromoplatinate(IV) hexahydrate

Catalog No. p-7080445
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8896603 1 g
AA8896606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8896603 Supplier Thermo Scientific Chemicals Supplier No. 08896603
Only null left

CAS: 39277-13-9 | Br6Na2Pt | 720.488 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39277-13-9
Molecular Formula Br6Na2Pt
Molecular Weight (g/mol) 720.488
MDL Number MFCD00014237
InChI Key RFZKZIKSQSDBBI-UHFFFAOYSA-H
Synonym disodium hexabromoplatinate, sodium hexabromoplatinate iv, disodium hexabromoplatinumdiuide, platinate 2-, hexabromo-, disodium, oc-6-11
PubChem CID 20474473
IUPAC Name disodium;hexabromoplatinum(2-)
SMILES [Na+].[Na+].Br[Pt-2](Br)(Br)(Br)(Br)Br

Specifications

Physical Form Crystalline
Quantity 1 g
Formula Weight 828.49 (720.49 Anhydrous)
Chemical Name or Material Sodium hexabromoplatinate (IV) hexahydrate
Hazard Category H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
EINECSNumber 254-398-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only