missing translation for 'onlineSavingsMsg'
Learn More

Sodium glycolate, 97%

Catalog No. p-192526
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC351570250 25 g Glass bottle
AC351571000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC351570250 Supplier Thermo Scientific Chemicals Supplier No. 351570250
Only null left

CAS: 2836-32-0 | C2H3NaO3 | 98.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2836-32-0
Molecular Formula C2H3NaO3
Molecular Weight (g/mol) 98.03
MDL Number MFCD00065358
InChI Key VILMUCRZVVVJCA-UHFFFAOYSA-M
Synonym sodium glycolate, sodium hydroxyacetate, sodium 2-hydroxyacetate, sodium lycolate, monosodium glycolate, glycolic acid sodium salt, natriumglykolat, sodium glycollate, acetic acid, hydroxy-, monosodium salt, glycolic acid, monosodium salt
PubChem CID 517347
IUPAC Name sodium;2-hydroxyacetate
SMILES C(C(=O)[O-])O.[Na+]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NaOOCCH2OH
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 98.03
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Sodium glycolate, 97%
Show More Show Less
EINECSNumber 220-624-9
RTECSNumber MC5525000
TSCA TSCA

RUO – Research Use Only