missing translation for 'onlineSavingsMsg'
Learn More

Sodium Gluconate, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18606513
Change view
Click to view available options
Quantity:
12 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606513 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606513 Supplier Spectrum Chemical Mfg Cor Supplier No. S128612KG
Get Up to $500 in Chemicals with Purchase
View all promotions

S1286, 527-07-1, C6H11NaO7

Spectrum™ Chemical Sodium Gluconate, USP is used as surface cleaning agent for metals and glass bottles. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 527-07-1
Molecular Formula C6H11NaO7
Molecular Weight (g/mol) 218.14
InChI Key UPMFZISCCZSDND-UHFFFAOYNA-M
IUPAC Name sodium 2,3,4,5,6-pentahydroxyhexanoate
SMILES [Na+].OCC(O)C(O)C(O)C(O)C([O-])=O

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 12 kg
Percent Purity 98 to 102%
Grade USP