missing translation for 'onlineSavingsMsg'
Learn More

Sodium Gluconate, USP, 98-102%, Spectrum™ Chemical

Catalog No. p-7158506
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
18606511 500 g Poly Bottle
18606512 2.5 kg Poly Bottle
2 options

Catalog No. 18606511

Supplier: Spectrum Chemical Mfg Cor S1286500GM

Only null left

S1286, 527-07-1, C6H11NaO7

Spectrum™ Chemical Sodium Gluconate, USP is used as surface cleaning agent for metals and glass bottles. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 527-07-1
Molecular Formula C6H11NaO7
Molecular Weight (g/mol) 218.14
InChI Key UPMFZISCCZSDND-UHFFFAOYNA-M
IUPAC Name sodium 2,3,4,5,6-pentahydroxyhexanoate
SMILES [Na+].OCC(O)C(O)C(O)C(O)C([O-])=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Percent Purity 98 to 102%
Grade USP