missing translation for 'onlineSavingsMsg'
Learn More

Sodium Formate, Crystal, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1285500GM

 View more versions of this product

Catalog No. 18613496


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

141-53-7
500 g
ACS
1
99%
Poly Bottle

Chemical Identifiers

141-53-7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.