missing translation for 'onlineSavingsMsg'
Learn More

Sodium formate, ACS reagent

Catalog No. AC446951000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446951000 100 g Plastic bottle
AC446955000 500 g Plastic bottle
AC446950025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC446951000 Supplier Thermo Scientific Chemicals Supplier No. 446951000

CAS: 141-53-7 | CHNaO2 | 68.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.01
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
PubChem CID 2723810
ChEBI CHEBI:62965
IUPAC Name sodium;formate
SMILES C(=O)[O-].[Na+]

Specifications

Melting Point 258.0°C to 261.0°C
Boiling Point 360.0°C
Packaging Plastic bottle
Linear Formula HCO2Na
Quantity 100 g
Merck Index 158756
Solubility Information Solubility in water: 550g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,insoluble in ether
Formula Weight 68.01
Grade ACS Reagent
Insoluble Matter 0.005% max
Chemical Name or Material Sodium formate
Show More Show Less
EINECSNumber 205-488-

RUO – Research Use Only