missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium formate, 99%
CAS: 141-53-7 | CHNaO2 | 68.007 g/mol
$102.46 - $102.46
Chemical Identifiers
| CAS | 141-53-7 |
|---|---|
| Molecular Formula | CHNaO2 |
| Molecular Weight (g/mol) | 68.007 |
| MDL Number | MFCD00013101 |
| InChI Key | HLBBKKJFGFRGMU-UHFFFAOYSA-M |
| Synonym | sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744 |
| PubChem CID | 2723810 |
| ChEBI | CHEBI:62965 |
| IUPAC Name | sodium;formate |
| SMILES | C(=O)[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC148230010
|
Thermo Scientific Chemicals
148230010 |
1 kg | Plastic bottle |
Each for $102.46
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 141-53-7 | |
| 68.007 | |
| HLBBKKJFGFRGMU-UHFFFAOYSA-M | |
| 2723810 | |
| sodium;formate |
| CHNaO2 | |
| MFCD00013101 | |
| sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744 | |
| CHEBI:62965 | |
| C(=O)[O-].[Na+] |
Specifications
| 141-53-7 | |
| White | |
| Authentic | |
| Plastic bottle | |
| HCO2Na | |
| 1 kg | |
| 8458 | |
| sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744 | |
| HLBBKKJFGFRGMU-UHFFFAOYSA-M | |
| sodium;formate | |
| 2723810 | |
| 68.01 | |
| Crystalline Powder |
| 258.0°C to 261.0°C | |
| 360.0°C | |
| 0.99 | |
| CHNaO2 | |
| MFCD00013101 | |
| 02, 8 | |
| 15, 8756 | |
| Solubility in water: 550g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol, insoluble in ether | |
| C(=O)[O-].[Na+] | |
| 68.007 | |
| CHEBI:62965 | |
| 0.99 | |
| Sodium formate |
RUO – Research Use Only