missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Formate, ≥99%, ACS Reagent, Honeywell Fluka™
ACS Reagent,≥99.0%
$320.47 - $320.47
Chemical Identifiers
| CAS | 141-53-7 |
|---|---|
| Molecular Formula | CHNaO2 |
| Molecular Weight (g/mol) | 68.007 |
| MDL Number | MFCD00013101 |
| InChI Key | HLBBKKJFGFRGMU-UHFFFAOYSA-M |
| Synonym | sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744 |
| PubChem CID | 2723810 |
| ChEBI | CHEBI:62965 |
| IUPAC Name | sodium;formate |
| SMILES | C(=O)[O-].[Na+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 141-53-7 | |
| 68.007 | |
| HLBBKKJFGFRGMU-UHFFFAOYSA-M | |
| 2723810 | |
| sodium;formate |
| CHNaO2 | |
| MFCD00013101 | |
| sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744 | |
| CHEBI:62965 | |
| C(=O)[O-].[Na+] |
Specifications
| 259°C to 262°C | |
| White | |
| CHNaO2 | |
| MFCD00013101 | |
| 3595134 | |
| Soluble in water | |
| C(=O)[O-].[Na+] | |
| 68.007 | |
| CHEBI:62965 | |
| ≥99.0% | |
| Poly Bottle | |
| 1.92g/cm3 |
| 141-53-7 | |
| 500 g | |
| HCOONa | |
| NONH for all modes of transport | |
| sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744 | |
| HLBBKKJFGFRGMU-UHFFFAOYSA-M | |
| sodium;formate | |
| 2723810 | |
| 68.01g/mol | |
| ACS Reagent | |
| Powder or crystals | |
| Sodium Formate |
Safety and Handling
EINECSNumber : 205-488-0
RTECSNumber : LR0350000