Learn More
Sodium formate, 98%
CAS: 141-53-7 | CHNaO2 | 68.007 g/mol
Supplier: Thermo Scientific Chemicals A1781330
Description
Sodium formate is used in analytical chemistry to precipitate noble metals. It is used for tanning leather, for dyeing and printing fabrics. It acts as an electroplating agent, an acidulant in the textile industry, a reducing agent, a mordant, a complexing agent. Further, it is used in photographic fixing baths. In addition, it is used as a buffering agent for strong mineral acids and food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium formate is used in analytical chemistry to precipitate noble metals. It is used for tanning leather, for dyeing and printing fabrics. It acts as an electroplating agent, an acidulant in the textile industry, a reducing agent, a mordant, a complexing agent. Further, it is used in photographic fixing baths. In addition, it is used as a buffering agent for strong mineral acids and food additive.
Solubility
Soluble in water, alcohol, glycerol and formic acid. Insoluble in ether.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids. Moisture sensitive
Spécifications
| Sodium formate | |
| 259°C to 262°C | |
| CHNaO2 | |
| 250 g | |
| Hygroscopic | |
| sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744 | |
| HLBBKKJFGFRGMU-UHFFFAOYSA-M | |
| sodium;formate | |
| 2723810 | |
| 68.01 |
| 141-53-7 | |
| 1.92 | |
| MFCD00013101 | |
| 3595134 | |
| 14,8621 | |
| Soluble in water,alcohol,glycerol and formic acid. Insoluble in ether. | |
| C(=O)[O-].[Na+] | |
| 68.007 | |
| CHEBI:62965 | |
| 98% |
Identifiants chimiques
| 141-53-7 | |
| 68.007 | |
| HLBBKKJFGFRGMU-UHFFFAOYSA-M | |
| 2723810 | |
| sodium;formate |
| CHNaO2 | |
| MFCD00013101 | |
| sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744 | |
| CHEBI:62965 | |
| C(=O)[O-].[Na+] |
Sécurité et manipulation
P210-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H320-H335-H500
missing translation for 'einecsNumber' : 205-488-0
missing translation for 'rtecsNumber' : LR0350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only