missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™

Catalog No. 6001386
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001386 1 kg
60045804 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001386 Supplier Honeywell Chemicals Supplier No. 301051KG
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
SMILES [F-].[Na+]

Specifications

Quantity 1 kg
Linear Formula NaF
UN Number 1690
IUPAC Name sodium;fluoride
Formula Weight 41.99g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Vapor Pressure 1.4mmHg (0°C)
Chemical Name or Material Sodium Fluoride, Reag. Ph. Eur. reag. ISO
Hazard Statement P305 + P351 + P338
Precautionary Statement H301-H315-H319
EINECSNumber 231-667-8
RTECSNumber WB0350000