missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride, puriss., Honeywell Fluka™

Catalog No. 6001391
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001391 1 kg
60044462 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001391 Supplier Honeywell Chemicals Supplier No. 011481KG
Only null left

Puriss., meets analytical specification of BP, Ph. Eur., USP, 98.5-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
SMILES [F-].[Na+]

Specifications

Quantity 1 kg
Linear Formula NaF
UN Number 1690
IUPAC Name sodium fluoride
Formula Weight 41.99g/mol
Percent Purity 98.5 to 100.5% (calc. to the dried substance)
Grade puriss.
Vapor Pressure 1.4mmHg (0°C)
Chemical Name or Material Sodium Fluoride, Meets analytical specification of Ph. Eur. BP, USP
Hazard Statement P305 + P351 + P338
Precautionary Statement H301-H315-H319
EINECSNumber 231-667-8
RTECSNumber WB0350000