missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Fluoride (Powder/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical S2993
Specifications
| 1700°C | |
| 993°C | |
| White | |
| Powder/Solid | |
| ≥99 % | |
| MFCD00003524 | |
| PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
| sodium fluoride | |
| 5235 | |
| 41.99 | |
| Certified ACS | |
| 0.3% max. | |
| 1mmHg at 1077°C |
| Sodium Fluoride | |
| 7681-49-4 | |
| 7.4 | |
| 3 kg | |
| FNa | |
| sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril | |
| [F-].[Na+] | |
| 41.99 | |
| CHEBI:28741 | |
| ≥99% | |
| Pass Test | |
| Poly Bottle |
Chemical Identifiers
| 7681-49-4 | |
| 41.99 | |
| PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
| 5235 | |
| sodium fluoride |
| FNa | |
| MFCD00003524 | |
| sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril | |
| CHEBI:28741 | |
| [F-].[Na+] |
Safety and Handling
DOTInformation : DOT Class 6.1, : Poison