missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoride (Powder/Certified ACS), Fisher Chemical™

Catalog No. S299100
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S299100 100 g Poly Bottle
S299500 500 g Poly Bottle
S2993 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S299100 Supplier Thermo Fisher Scientific Supplier No. S299 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Boiling Point 1700°C
Melting Point 993°C
Color White
pH 7.4
Physical Form Powder/Solid
Quantity 100 g
Assay Percent Range ≥99 %
Formula Weight 41.99
Percent Purity ≥99%
Grade Certified ACS
Identification Pass Test
Loss on Drying 0.3% max.
Packaging Poly Bottle
Vapor Pressure 1mmHg at 1077°C
Chemical Name or Material Sodium Fluoride
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison