Learn More
Sodium fluoride, Optical Grade, Thermo Scientific Chemicals
$457.22 - $457.22
Chemical Identifiers
| CAS | 7681-49-4 |
|---|---|
| Molecular Formula | FNa |
| Molecular Weight (g/mol) | 41.99 |
| MDL Number | MFCD00003524 |
| InChI Key | PUZPDOWCWNUUKD-UHFFFAOYSA-M |
| Synonym | sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril |
| PubChem CID | 5235 |
| ChEBI | CHEBI:28741 |
| SMILES | [F-].[Na+] |
Description
It has medical and other applications including complexing of magnesium ions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7681-49-4 | |
| 41.99 | |
| PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
| 5235 | |
| [F-].[Na+] |
| FNa | |
| MFCD00003524 | |
| sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril | |
| CHEBI:28741 |
Specifications
| 1704°C | |
| 7681-49-4 | |
| Broken Crystals | |
| FNa | |
| sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril | |
| PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
| sodium fluoride | |
| 5235 | |
| 41.99 | |
| Odorless | |
| White crystals | |
| Sodium fluoride |
| 993°C | |
| White | |
| 50 g | |
| MFCD00003524 | |
| Soluble in water. Insoluble in alcohol. | |
| [F-].[Na+] | |
| 41.99 | |
| CHEBI:28741 | |
| Optical | |
| 1.336 | |
| 2.78 |
Safety and Handling
Do not breathe dust. Wear suitable protective clothing. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H301-H315-H319
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 231-667-8
RTECSNumber : WB0350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only