missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, ACS, 99% min, Thermo Scientific Chemicals

Catalog No. AA11561A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11561A1 1 kg
AA1156130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA11561A1 Supplier Thermo Scientific Chemicals Supplier No. 011561A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Steel Degreasing Agent, in Tin Electroplating, Flux

It is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including it use in synthetic chemistry, for example in Finkelstein reaction for preparating fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Boiling Point 1704°C
Melting Point 993°C
Color White
Physical Form Powder
Quantity 1 kg
Odor Odorless
Refractive Index 1.336
Appearance White powder
Vapor Pressure 1.3 hPa (1mm Hg)
Assay Percent Range 99% min.
Density 2.78
Solubility Information Slightly Soluble in water. Insoluble in alcohol
Formula Weight 41.99
Percent Purity ≥99%
Grade ACS
Chemical Name or Material Sodium fluoride, For ACS analysis
Show More Show Less
Hazard Category H301-H315-H319
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 231-667-8
RTECSNumber WB0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only