missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A130190I

 View more versions of this product

Catalog No. AAA130190I


Only null left
Explore available promotions

Description

Description

Sodium fluoride is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including it use in synthetic chemistry, for example in Finkelstein reaction for preparating fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium fluoride is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including it use in synthetic chemistry, for example in Finkelstein reaction for preparating fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.

Solubility
Slightly soluble in water. Insoluble in alcohol

Notes
It must be kept away from oxidizing agents, metals, acids, alkalis. It is less hygroscopic.
Specifications

Specifications

1704°C
993°C
5000 g
NaF
UN1690
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride
5235
41.99
Odorless
2.78
Sodium fluoride
7681-49-4
FNa
MFCD00003524
14,8618
Slightly soluble in water. Insoluble in alcohol
[F-].[Na+]
41.99
CHEBI:28741
99%
1.336

Chemical Identifiers

7681-49-4
41.99
PUZPDOWCWNUUKD-UHFFFAOYSA-M
5235
[F-].[Na+]
FNa
MFCD00003524
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
CHEBI:28741
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H301-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SODIUM FLUORIDE, SOLID

EINECSNumber : 231-667-8

RTECSNumber : WB0350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only