missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, ≥99%, ACS reagent, powder, Thermo Scientific Chemicals

Catalog No. AC424320050
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424320050 5 g Plastic bottle
AC424325000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC424320050 Supplier Thermo Scientific Chemicals Supplier No. 424320050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
SMILES [F-].[Na+]

Specifications

Boiling Point 1700.0°C
Melting Point 993.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range ≥99%
Linear Formula NaF
Fieser 01,1073; 02,382
Merck Index 15, 8754
Solubility Information Solubility in water: 40g/L (25°C). Other solubilities: slightly soluble in alcohol
IUPAC Name sodium;fluoride
Formula Weight 41.98
Percent Purity ≥99%
Grade ACS Reagent
Loss on Drying 0.3% max. (150°C)
Packaging Plastic bottle
Chemical Name or Material Sodium fluoride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs
EINECSNumber 231-667-8
RTECSNumber WB0350000
TSCA TSCA

RUO – Research Use Only