missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium fluoride, 97%, extra pure, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
| Boiling Point | 1700.0°C |
| Melting Point | 993.0°C |
| Color | White |
| Physical Form | Crystals or Powder |
| Quantity | 500 g |
| Assay Percent Range | 97% |
| Linear Formula | NaF |
| Fieser | 01,1073; 02,382 |
| Merck Index | 15, 8754 |
| Solubility Information | Solubility in water: 40g/L (25°C). Other solubilities: slightly soluble in alcohol |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. Contact with acids liberates very toxic gas. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. If skin irritation occurs |
| EINECSNumber | 231-667-8 |
| RTECSNumber | WB0350000 |
| TSCA | TSCA |
RUO – Research Use Only