missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, 200mM aq. soln., Thermo Scientific Chemicals

Catalog No. AAJ60251AK
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60251AK 250 mL
AAJ60251AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60251AK Supplier Thermo Scientific Chemicals Supplier No. J60251AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Linear Formula NaF
Formula Weight 41.99
Grade Ungraded
Concentration or Composition (by Analyte or Components) 200mM aq. soln.
Chemical Name or Material Sodium fluoride
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only