Learn More
Sodium ethoxide, 96%
CAS: 141-52-6 | C2H5NaO | 68.051 g/mol
Supplier: Thermo Scientific Chemicals L0623036
Description
Sodium ethoxide is used as a strong base in organic synthesis. It is actively involved in the Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether. It is widely used as a base for the generation of carbanions which are utilized for alkylation and condensation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium ethoxide is used as a strong base in organic synthesis. It is actively involved in the Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether. It is widely used as a base for the generation of carbanions which are utilized for alkylation and condensation reactions.
Solubility
Soluble in ethanol and methanol.
Notes
Moisture sensitive. It undergoes hydrolysis rapidly to form sodium hydroxide in the presence of moist air. Incompatible with water, strong bases, acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents, peroxides, acids and chlorinated solvents.
Specifications
| Sodium ethoxide | |
| >300°C | |
| 30°C (86°F) | |
| C2H5NaO | |
| 500 g | |
| 3593646 | |
| 14,8613 | |
| Soluble in ethanol and methanol. | |
| CC[O-].[Na+] | |
| 68.051 | |
| CHEBI:52096 | |
| 96% |
| 141-52-6 | |
| 13 | |
| Irritating | |
| MFCD00012417 | |
| UN3206 | |
| Air and moisture sensitive | |
| sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1 | |
| QDRKDTQENPPHOJ-UHFFFAOYSA-N | |
| sodium;ethanolate | |
| 2723922 | |
| 68.05 |
Chemical Identifiers
| 141-52-6 | |
| 68.051 | |
| QDRKDTQENPPHOJ-UHFFFAOYSA-N | |
| 2723922 | |
| sodium;ethanolate |
| C2H5NaO | |
| MFCD00012417 | |
| sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1 | |
| CHEBI:52096 | |
| CC[O-].[Na+] |
Safety and Handling
GHS H Statement
H251-H314-H318
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P235+P410-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H314-H335-H500-X
DOTInformation : Hazard Class: 8; Packaging Group: I
EINECSNumber : 205-487-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only