missing translation for 'onlineSavingsMsg'
Learn More

Sodium ethoxide, 96%

CAS: 141-52-6 | C2H5NaO | 68.051 g/mol

Supplier:  Thermo Scientific Chemicals L062300P

 View more versions of this product

Catalog No. AAL062300P

Only null left
Explore available promotions

Description

Description

Sodium ethoxide is used as a strong base in organic synthesis. It is actively involved in the Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether. It is widely used as a base for the generation of carbanions which are utilized for alkylation and condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium ethoxide is used as a strong base in organic synthesis. It is actively involved in the Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether. It is widely used as a base for the generation of carbanions which are utilized for alkylation and condensation reactions.

Solubility
Soluble in ethanol and methanol.

Notes
Moisture sensitive. It undergoes hydrolysis rapidly to form sodium hydroxide in the presence of moist air. Incompatible with water, strong bases, acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents, peroxides, acids and chlorinated solvents.
Specifications

Specifications

Sodium ethoxide
>300°C
30°C (86°F)
C2H5NaO
2000 g
3593646
14,8613
Soluble in ethanol and methanol.
CC[O-].[Na+]
68.051
CHEBI:52096
96%
141-52-6
13
Irritating
MFCD00012417
UN3206
Air and moisture sensitive
sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
QDRKDTQENPPHOJ-UHFFFAOYSA-N
sodium;ethanolate
2723922
68.05

Chemical Identifiers

141-52-6
68.051
QDRKDTQENPPHOJ-UHFFFAOYSA-N
2723922
sodium;ethanolate
C2H5NaO
MFCD00012417
sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
CHEBI:52096
CC[O-].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H251-H314-H318
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P235+P410-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H314-H335-H500-X

DOTInformation : Hazard Class: 8; Packaging Group: I

EINECSNumber : 205-487-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only