missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecyl Sulfate, MP Biomedicals
$508.38 - $508.38
Specifications
| Chemical Name or Material | SODIUM DODECYL SULFATE |
|---|---|
| Color | White |
| Physical Form | Powder |
| Melting Point | 180°C to 210°C |
| Molecular Formula | C12H25NaO4S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
MP04811032
|
MP Biomedicals Inc
04811032 |
500 g |
Each for $508.38
|
|
|||||
Description
- Used to solubilize and denature proteins for denaturing-PAGE
- Most proteins bind SDS in a ratio of 1.4 g SDS per gram of protein
- The charges intrinsic to the protein become insignificant compared to the overall negative charge provided by the bound SDS
- The charge to mass ratio is essentially the same for each protein and will migrate in the gel based only on their size
Specifications
| SODIUM DODECYL SULFATE | |
| Powder | |
| C12H25NaO4S | |
| MFCD00036175 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| 288.38 | |
| CHEBI:8984 | |
| ≥99% |
| White | |
| 180°C to 210°C | |
| C12H25O4SNa | |
| Soluble in water (200mg/mL - clear, faint yellow solution), and ethanol (0.1g/10 mL). | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 3423265 | |
| 288.38 | |
| Ultra Pure |