missing translation for 'onlineSavingsMsg'
Learn More

Sodium DL-Mandelate 99.0+%, TCI America™

Catalog No. M004125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M004125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M004125G Supplier TCI America Supplier No. M004125G

Chemical Identifiers

CAS 114-21-6
Molecular Formula C8H7NaO3
Molecular Weight (g/mol) 174.131
MDL Number MFCD00051122
InChI Key FWDLHTBMGQEUDU-UHFFFAOYSA-M
Synonym DL-Mandelic Acid Sodium Salt
PubChem CID 23675514
IUPAC Name sodium;2-hydroxy-2-phenylacetate
SMILES C1=CC=C(C=C1)C(C(=O)[O-])O.[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 174.13
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium DL-Mandelate