missing translation for 'onlineSavingsMsg'
Learn More

Sodium DL-Lactate Solution, USP, 60%, Spectrum™ Chemical

Catalog No. p-7158213
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606491 4 L Amber Glass Bottle
18606492 20 L Poly Drum
18606493 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606491 Supplier Spectrum Chemical Mfg Cor Supplier No. SO1794LT
Only null left

SO179, 72-17-3, C3H5NaO3

Spectrum™ Chemical Sodium DL-Lactate Solution, 60 Percent, USP is used for the restoration of extracellular fluid and electrolytes balances. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

Molecular Formula C3H5NaO3
Molecular Weight (g/mol) 112.06
InChI Key NGSFWBMYFKHRBD-UHFFFAOYNA-M
IUPAC Name sodium 2-hydroxypropanoate
SMILES [Na+].CC(O)C([O-])=O

Specifications

pH 5.0 to 9.0
Packaging Amber Glass Bottle
Quantity 4 L
Percent Purity 58.8 to 61.2%
Grade USP