missing translation for 'onlineSavingsMsg'
Learn More

Sodium DL-lactate, 60% w/w aq. soln.

Catalog No. p-7051557
Change view
Click to view available options
Quantity:
250 mL
1 L
Catalog No. Quantity
AA41529AK 250 mL
AA41529K2 1 L
2 options

Catalog No. AA41529AK

Supplier: Thermo Scientific Chemicals 041529AK

Only null left

Useful chiral synthon; building block for depsipeptides | CAS: 72-17-3 | C3H5NaO3 | 112.06 g/mol

Useful chiral synthon; building block for depsipeptides.Sodium DL-lactate, 60% w/w aq. Soln is used as a food additive, plasticizer for casein, preservative, acidity regulator, bulking agent, and in shampoo and liquid soaps. It is also used for the treatment for arrhythmias and sympathomimetics. It is useful chiral synthon and building block for depsipeptides. It is used as a corrosion inhibitor in alcohol antifreeze mixtures, flavoring agent, emulsifier, corrosion inhibitor and pH control agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72-17-3
Molecular Formula C3H5NaO3
Molecular Weight (g/mol) 112.06
MDL Number MFCD00065400
InChI Key NGSFWBMYFKHRBD-UHFFFAOYSA-M
Synonym sodium lactate, lactic acid sodium salt, monosodium lactate, sodium dl-lactate, lacolin, per-glycerin, lactic acid, monosodium salt, sodium alpha-hydroxypropionate, monosodium 2-hydroxypropanoate, propanoic acid, 2-hydroxy-, monosodium salt
PubChem CID 23666456
ChEBI CHEBI:75228
IUPAC Name sodium;2-hydroxypropanoate
SMILES CC(C(=O)[O-])O.[Na+]

Specifications

Color Colorless
Density 1.326 g/mL
Odor Odorless
Linear Formula C3H5NaO3
Quantity 250 mL
Beilstein 4332999
Merck Index 14,8635
Solubility Information Miscible with water and alcohol.
Formula Weight 112.06
Concentration or Composition (by Analyte or Components) 60% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Sodium DL-lactate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 200-772-0
RTECSNumber OD5680000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only