missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dithionite (Sodium Hydrosulfite), Reagents

Sodium Dithionite (Sodium Hydrosulfite), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

$140.60 - $5,168.47

Chemical Identifiers

CAS 7775-14-6
Molecular Formula Na2O4S2
Molecular Weight (g/mol) 174.10
InChI Key JVBXVOWTABLYPX-UHFFFAOYSA-L
Synonym Sodium Hydrosulfite
IUPAC Name disodium sulfinatosulfinate
SMILES [Na+].[Na+].[O-]S(=O)S([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-337-024
View Documents
Labchem Inc
C2316300100KG
100 kg N/A
Only null left
 
01-337-023
View Documents
Labchem Inc
C231630025A5
2.5 kg N/A
Only null left
 
01-337-022
View Documents
Labchem Inc
C2316300500C2
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7775-14-6
174.10
Sodium Hydrosulfite
[Na+].[Na+].[O-]S(=O)S([O-])=O
Na2O4S2
JVBXVOWTABLYPX-UHFFFAOYSA-L
disodium sulfinatosulfinate

Specifications

>80°C
White to Off-White
8 to 10.5
100 kg
Sodium Hydrosulfite
[Na+].[Na+].[O-]S(=O)S([O-])=O
174.10
Steel Drum
Sodium Dithionite (Sodium Hydrosulfite)
52°C
7775-14-6
Solid
Na2O4S2
JVBXVOWTABLYPX-UHFFFAOYSA-L
disodium sulfinatosulfinate
Pure
2.19
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'