missing translation for 'onlineSavingsMsg'
Learn More

Sodium Diphenylaminesulfonate, ACS Reagent, Reagents

Catalog No. p-200021029
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337733 25 g
01337732 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337733 Supplier Labchem Inc Supplier No. C131610025C1
Only null left

Sodium Diphenylaminesulfonate, ACS Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6152-67-6
Molecular Formula C12H11NNaO3S
Molecular Weight (g/mol) 272.27
InChI Key VMVSDVIYIMNZBS-UHFFFAOYSA-N
Synonym Diphenylaminesulfonic Acid Sodium Salt
IUPAC Name 4-(phenylamino)benzene-1-sulfonic acid sodium
SMILES [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Color Off-White
Physical Form Solid
Quantity 25 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1
Chemical Name or Material Sodium Diphenylaminesulfonate, ACS Reagent