missing translation for 'onlineSavingsMsg'
Learn More

Sodium diphenylamine-4-sulfonate, 98%

CAS: 6152-67-6 | C12H11NNaO3S | 272.27 g/mol

Supplier:  Thermo Scientific Chemicals A1976814

 View more versions of this product

Catalog No. AAA1976814


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium diphenylamine-4-sulfonate
C12H11NNaO3S
25 g
VMVSDVIYIMNZBS-UHFFFAOYSA-N
sodium;4-anilinobenzenesulfonate
517002
98%
6152-67-6
MFCD00036468
sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
272.27
271.27

Chemical Identifiers

6152-67-6
272.27
VMVSDVIYIMNZBS-UHFFFAOYSA-N
517002
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
C12H11NNaO3S
MFCD00036468
sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
sodium;4-anilinobenzenesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 228-165-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only