missing translation for 'onlineSavingsMsg'
Learn More

Sodium Diformylamide 97.0+%, TCI America™

Catalog No. D247925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D247925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D247925G Supplier TCI America Supplier No. D247925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18197-26-7
Molecular Formula C2H2NNaO2
Molecular Weight (g/mol) 95.033
MDL Number MFCD00145302
InChI Key QJXDSDLNUKLDBP-UHFFFAOYSA-M
Synonym sodium diformylamide, diformylamide sodium salt, diformylimide sodium salt, sodium diformylazanide, sodium diformamide, diformylamino sodium, diformylsodium, acmc-209eii, sodium dimethanoylazanide, ksc538a2h
PubChem CID 10877056
IUPAC Name sodium;diformylazanide
SMILES C(=O)[N-]C=O.[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 95.03
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Diformylamide
TSCA No
Recommended Storage Refrigerator