missing translation for 'onlineSavingsMsg'
Learn More

Sodium diformamide, 98%, Thermo Scientific™

Chemical Identifiers

CAS 18197-26-7
Molecular Formula C2H2NNaO2
Molecular Weight (g/mol) 95.03
MDL Number MFCD00145302
InChI Key QJXDSDLNUKLDBP-UHFFFAOYSA-M
Synonym sodium diformylamide, diformylamide sodium salt, diformylimide sodium salt, sodium diformylazanide, sodium diformamide, diformylamino sodium, diformylsodium, acmc-209eii, sodium dimethanoylazanide, ksc538a2h
PubChem CID 10877056
IUPAC Name sodium;diformylazanide
SMILES C(=O)[N-]C=O.[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC380430250
SDS View Documents
Thermo Scientific Chemicals
380430250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

18197-26-7
95.03
QJXDSDLNUKLDBP-UHFFFAOYSA-M
10877056
C(=O)[N-]C=O.[Na+]
C2H2NNaO2
MFCD00145302
sodium diformylamide, diformylamide sodium salt, diformylimide sodium salt, sodium diformylazanide, sodium diformamide, diformylamino sodium, diformylsodium, acmc-209eii, sodium dimethanoylazanide, ksc538a2h
sodium;diformylazanide

Specifications

18197-26-7
98%
NaN(COH)2
sodium diformylamide, diformylamide sodium salt, diformylimide sodium salt, sodium diformylazanide, sodium diformamide, diformylamino sodium, diformylsodium, acmc-209eii, sodium dimethanoylazanide, ksc538a2h
C(=O)[N-]C=O.[Na+]
95.03
95.03
Authentic
Sodium diformamide
25g
C2H2NNaO2
MFCD00145302
QJXDSDLNUKLDBP-UHFFFAOYSA-M
sodium;diformylazanide
10877056
98%
Glass bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only