missing translation for 'onlineSavingsMsg'
Learn More

Sodium diethyldithiocarbamate trihydrate, ACS reagent, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. p-7187753
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018982 100 g
111018177 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018982 Supplier Merck Emd Millipore Supplier No. 71480100G
Only null left

Sodium diethyldithiocarbamate trihydrate is a chelating agent, generally used in mobilizing toxic metals from the human tissues and experimental animals. It exists as a white or colorless solid, and is freely soluble with gradual decomposition.

Chemical Identifiers

MDL Number MFCD00150617
Synonym Cupral; Diethyldithiocarbamic acid sodium salt

Specifications

Melting Point 95°C to 98.5°C
CAS 20624-25-3
Quantity 100 g
Linear Formula (C2H5)2 NCSSNa · 3H2 O
Formula Weight 225.31
Percent Purity ≥99.0%
Grade ACS Reagent
Density Vapor Density: 5.9 (vs. air)
RTECSNumber EZ6550000