missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium diethyldithiocarbamate trihydrate, ACS reagent, ≥99.0%, MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | 95°C to 98.5°C |
| CAS | 20624-25-3 |
| Quantity | 500 g |
| Linear Formula | (C2H5)2 NCSSNa · 3H2 O |
| Formula Weight | 225.31 |
| Percent Purity | ≥99.0% |
| Grade | ACS Reagent |
| Density | Vapor Density: 5.9 (vs. air) |
Safety and Handling
| RTECSNumber | EZ6550000 |