Learn More
Sodium dicyanamide, 97%, pure, Thermo Scientific™
$99.97 - $1,081.52
Chemical Identifiers
| CAS | 1934-75-4 |
|---|---|
| Molecular Formula | C2N3Na |
| Molecular Weight (g/mol) | 89.03 |
| MDL Number | MFCD00003535 |
| InChI Key | IXBPPZBJIFNGJJ-UHFFFAOYSA-N |
| Synonym | sodium dicyanamide, sodium dicyanoazanide, sodium dicyanoamide, unii-i9c96i1mre, cyanamide, cyano-, sodium salt, cyanamide,cyano-,sodium salt, i9c96i1mre, cyanamide, n-cyano-, sodium salt 1:1, dicyanamide, sodium salt, nadca |
| PubChem CID | 4350929 |
| IUPAC Name | sodium;dicyanoazanide |
| SMILES | [Na+].N#C[N-]C#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC176340250
|
N/A | 25g | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC176341000
|
N/A | 100g | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC176345000
|
N/A | 500g | Plastic bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C2N3Na | |
| MFCD00003535 | |
| sodium dicyanamide, sodium dicyanoazanide, sodium dicyanoamide, unii-i9c96i1mre, cyanamide, cyano-, sodium salt, cyanamide,cyano-,sodium salt, i9c96i1mre, cyanamide, n-cyano-, sodium salt 1:1, dicyanamide, sodium salt, nadca | |
| sodium;dicyanoazanide |
Specifications
| 1934-75-4 | |
| 2% max. (NaCl) (Argentometry) | |
| NaN(CN)2 | |
| 03, III, 154 | |
| Solubility in water: 260g/L (30°C). Other solubilities: insoluble in aceton,etoh and ethyl acetate | |
| [Na+].N#C[N-]C#N | |
| 89.03 | |
| 89.03 | |
| Pure |
| Authentic | |
| C2N3Na | |
| MFCD00003535 | |
| sodium dicyanamide, sodium dicyanoazanide, sodium dicyanoamide, unii-i9c96i1mre, cyanamide, cyano-, sodium salt, cyanamide,cyano-,sodium salt, i9c96i1mre, cyanamide, n-cyano-, sodium salt 1:1, dicyanamide, sodium salt, nadca | |
| IXBPPZBJIFNGJJ-UHFFFAOYSA-N | |
| sodium;dicyanoazanide | |
| 4350929 | |
| 97% | |
| Sodium dicyanamide, Pure |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lens
GHS Signal Word: Warning
EINECSNumber : 217-703-5
RUO – Research Use Only