missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dichromate, Granular, Technical, Spectrum™ Chemical

Catalog No. 18613472
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613472 500 g Poly Bottle
18613470 125 g Poly Bottle
18613468 2.5 kg Poly Bottle
18613471 12 kg Poly Bottle
18613469 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613472 Supplier Spectrum Chemical Mfg Cor Supplier No. S1268500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7789-12-0

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 500 g
Molecular Weight (g/mol) 298.00
Grade Technical
Packaging Poly Bottle
Chemical Name or Material Sodium Dichromate, Dihydrate, Granular

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.