missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dichromate Dihydrate, Purified, Reagents

Catalog No. 01337026
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
50 lb.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337026 500 g
01337028 10 kg
01337027 2.5 kg
01337025 50 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337026 Supplier Labchem Inc Supplier No. C2316000500A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Dichromate Dihydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
Molecular Weight (g/mol) 298.00
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym Sodium Bichromate
IUPAC Name disodium [(oxidodioxochromio)oxy]chromiumoylolate dihydrate
SMILES O.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

Specifications

Boiling Point 400°C
Melting Point 357°C
Color Orange/Red
Physical Form Solid
Quantity 500 g
Percent Purity 102%
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.35
Chemical Name or Material Sodium Dichromate Dihydrate, Purified