missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dichromate Dihydrate, ACS Reagent, Reagents
Specifications
Specifications
| Boiling Point | 400°C |
| Melting Point | 357°C |
| Color | Orange/Red |
| Physical Form | Solid |
| Quantity | 500 g |
| Percent Purity | ≥99.5% |
| Grade | ACS |
| Packaging | Poly Bottle |
| Specific Gravity | 2.35 |
| Chemical Name or Material | Sodium Dichromate Dihydrate, ACS Reagent |