missing translation for 'onlineSavingsMsg'
Learn More

Sodium dichromate dihydrate, ACS, 99.5-100.5%

CAS: 7789-12-0 | Cr2H4Na2O9 | 297.995 g/mol

$99.22 - $783.79

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
Molecular Weight (g/mol) 297.995
MDL Number MFCD00149166
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
PubChem CID 108005
IUPAC Name disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
SMILES O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3669222
View Documents
Thermo Scientific Chemicals
03669222
100 g
Each for $99.22
Only null left
 
AA3669236
View Documents
Thermo Scientific Chemicals
03669236
500 g
Each for $252.44
Only null left
 
AA36692A3
View Documents
Thermo Scientific Chemicals
036692A3
2 kg
Each for $783.79
Only null left
 
Description

Description

It is used as a strong oxidizing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-12-0
297.995
JYDRNIYTFCBIFC-UHFFFAOYSA-N
108005
O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]
Cr2H4Na2O9
MFCD00149166
sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate

Specifications

100°C-2H2O
Red
100 g
Cr2H4Na2O9
sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
JYDRNIYTFCBIFC-UHFFFAOYSA-N
disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
108005
ACS
Red crystalline
Sodium dichromate dihydrate, For analysis ACS
7789-12-0
Crystalline
99.5-100.5%
MFCD00149166
Soluble in water. Insoluble in alcohol.
O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]
297.995
298.00 (261.97 Anhydrous)
Hygroscopic
2.438 g/cm3 at 20°C
Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c

H272-H301-H312-H314-H317-H330-H334-H335-H340-H350-H360FD-H372

DOTInformation : DOT Class: 5.1; Packing Group: II

EINECSNumber : 234-190-3

RTECSNumber : HX7750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only