missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium dichromate dihydrate, ACS, 99.5-100.5%
Description
It is used as a strong oxidizing agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100°C-2H2O |
| Color | Red |
| Physical Form | Crystalline |
| Quantity | 2 kg |
| Sensitivity | Hygroscopic |
| Appearance | Red crystalline |
| Assay Percent Range | 99.5-100.5% |
| Density | 2.438 g/cm3 at 20°C |
| Solubility Information | Soluble in water. Insoluble in alcohol. |
| Formula Weight | 298.00 (261.97 Anhydrous) |
| Show More |
Safety and Handling
| Hazard Category | H272-H301-H312-H314-H317-H330-H334-H335-H340-H350-H360FD-H372 |
| Precautionary Statement | P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 5.1; Packing Group: II |
| EINECSNumber | 234-190-3 |
| RTECSNumber | HX7750000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only