missing translation for 'onlineSavingsMsg'
Learn More

Sodium dichromate dihydrate, ACS, 99.5-100.5%

Catalog No. AA3669222
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3669222 100 g
AA3669236 500 g
AA36692A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3669222 Supplier Thermo Scientific Chemicals Supplier No. 03669222

CAS: 7789-12-0 | Cr2H4Na2O9 | 297.995 g/mol

It is used as a strong oxidizing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
Molecular Weight (g/mol) 297.995
MDL Number MFCD00149166
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
PubChem CID 108005
IUPAC Name disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
SMILES O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 100°C-2H2O
Color Red
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.5-100.5%
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 298.00 (261.97 Anhydrous)
Grade ACS
Sensitivity Hygroscopic
Appearance Red crystalline
Density 2.438 g/cm3 at 20°C
Chemical Name or Material Sodium dichromate dihydrate, For analysis ACS
Show More Show Less
Hazard Category H272-H301-H312-H314-H317-H330-H334-H335-H340-H350-H360FD-H372
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 5.1; Packing Group: II
EINECSNumber 234-190-3
RTECSNumber HX7750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only