missing translation for 'onlineSavingsMsg'
Learn More

Sodium dichromate dihydrate, 98%, Thermo Scientific Chemicals

Catalog No. p-7046545
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1489022 100 g
AAL1489036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1489022 Supplier Thermo Scientific Chemicals Supplier No. L1489022

Sodium dichromate is used as a strong oxidizing agent in organic synthesis. It is used in metal finishing as an aid of corrosion resistance, manufacture of inorganic chromate pigments and in the preparation of colored glass and ceramic glazes. It is used as a mordant for acidic dyes to improve their color-fast properties and as a corrosion inhibitor in undercoats and primers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dichromate is used as a strong oxidizing agent in organic synthesis. It is used in metal finishing as an aid of corrosion resistance, manufacture of inorganic chromate pigments and in the preparation of colored glass and ceramic glazes. It is used as a mordant for acidic dyes to improve their color-fast properties and as a corrosion inhibitor in undercoats and primers.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Keep away from sources of ignition, combustible materials. Do not ingest. Do not breathe dust.

Chemical Identifiers

CAS 7789-12-0
Molecular Formula Cr2H4Na2O9
Molecular Weight (g/mol) 297.995
MDL Number MFCD00149166
InChI Key JYDRNIYTFCBIFC-UHFFFAOYSA-N
Synonym sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903
PubChem CID 108005
IUPAC Name disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate
SMILES O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 100°C (in - 2H2O)
Quantity 100 g
Linear Formula Na2Cr2O7·2H2O
UN Number UN3087
Merck Index 14,8609
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 298.00 (261.97 Anhydrous)
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.348
Chemical Name or Material Sodium dichromate dihydrate
Show More Show Less
Hazard Category H272-H301-H312-H314-H317-H330-H334-H335-H340-H350-H360FD-H372
Hazard Statement GHS H Statement
H272-H301-H330-H334-H340-H350-H360-H372-H314-H312-H317
May intensify fire; oxidizer.
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
EINECSNumber 234-190-3
RTECSNumber HX7750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only