missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dichromate, Crystal, 99.5%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 50 kg |
| Molecular Weight (g/mol) | 298.00 |
| Percent Purity | 99.5% |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Chemical Name or Material | Sodium Dichromate, Crystal |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.