missing translation for 'onlineSavingsMsg'
Learn More

Sodium dehydroacetate, 97%

Catalog No. aab2106022
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2106022 100 g
AAB2106036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2106022 Supplier Thermo Scientific Chemicals Supplier No. B2106022

CAS: 4418-26-2 | C8H7NaO4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium dehydroacetate is used in the food industry as a preservative for fruits, vegetables, fruit juice, honey, beer, milk, protein and fat products. Its antimicrobial properties play a vital role in personal care products such as skin care, suntan, sunscreen, shaving, tooth paste, hair and nail care products, and eye and facial makeup. It is used as a plasticizer.

Solubility
Soluble in water, propylene glycol and methanol. Insoluble in acetone and benzene.

Notes
Incompatible strong oxidizing agents and acids.

Chemical Identifiers

CAS 4418-26-2
Molecular Formula C8H7NaO4
MDL Number MFCD00040583

Specifications

Melting Point ∼295°C (decomposition)
pH 8.8 to 9.3
Quantity 100 g
Synonym 3-Acetyl-4-hydroxy-6-methyl-2-pyrone sodium salt; Dehydroacetic acid sodium salt
Solubility Information Soluble in water,propylene glycol and methanol. Insoluble in acetone and benzene.
Formula Weight 190.13
Percent Purity 97%
Chemical Name or Material Sodium dehydroacetate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 224-580-1
RTECSNumber UP8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only