missing translation for 'onlineSavingsMsg'
Learn More

Sodium Decyl Sulfate 98.0+%, TCI America™

Catalog No. p-7136006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S09755G Supplier TCI America Supplier No. S09755G
Only null left

Chemical Identifiers

CAS 142-87-0
Molecular Formula C10H21NaO4S
Molecular Weight (g/mol) 260.324
MDL Number MFCD00041881
InChI Key XZTJQQLJJCXOLP-UHFFFAOYSA-M
Synonym sodium decyl sulfate, sodium n-decyl sulfate, decyl sodium sulfate, sulfuric acid, monodecyl ester, sodium salt, sodium decyl sulphate, unii-al92m833sy, n-decyl sodium sulfate, sodium n-decylsulphate, sulfuric acid, monodecyl ester, sodium salt 1:1, sulfuric acid, decyl ester, sodium salt
PubChem CID 23665771
IUPAC Name sodium;decyl sulfate
SMILES CCCCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 192°C
Color White
Quantity 5 g
Formula Weight 260.32
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Decyl Sulfate
EINECSNumber (2)-1679
RTECSNumber WS7625000
TSCA Yes