missing translation for 'onlineSavingsMsg'
Learn More

Sodium decyl sulfate, ≥99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013103
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013103 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013103 Supplier Merck Emd Millipore Supplier No. 7144310G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium decyl sulfate is an anionic surfactant. It is a sodium salt monodecyl ester of sulfuric acid. It also has application in textile, leather and other industries as emulsifier, wetting agent, dispersant, fiber lubricant, and synthetic fatliquor.

Chemical Identifiers

MDL Number MFCD00041881
Synonym Decyl sulfate sodium salt

Specifications

CAS 142-87-0
Linear Formula CH3(CH2)9 OSO3 Na
Quantity 10 g
Formula Weight 260.33
Percent Purity ≥99.0% (T)
Grade For ion pair chromatography
RTECSNumber WS7625000