missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sodium D-glycero-D-guloheptonate dihydrate, 98+%
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 1 kg |
| Formula Weight | 284.19 |
| Percent Purity | ≥99% |
| Grade | Ungraded |
| Chemical Name or Material | Sodium D-glycero-D-guloheptonate Dihydrate |
Safety and Handling
| ShelfLife | 5 Years |
| EINECSNumber | 235-849-8 |